(S)-2-(3,4-Dichlorophenyl)pent-4-enoic acid ≥94%
Supplier: Matrix Scientific
060060-1G
102551-560EA
300
USD
102551-560
(S)-2-(3,4-Dichlorophenyl)pent-4-enoic acid ≥94%
(S)-2-(3,4-Dichlorophenyl)pent-4-enoic acid
Hazard: Irritant
Formula:
C₁₁H₁₀Cl₂O₂ |
MDL Number:
MFCD19442216 CAS Number: 147643-57-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...