Piperidine-N-carbaldehyde
Supplier: Matrix Scientific
Synonyms:
N-Formylpiperidine
, 1-Formylpiperidine
058346-100G
058346-250G
102663-768EA
75.38
USD
102663-768
102663-770
Piperidine-N-carbaldehyde
Piperidine-N-carbaldehyde
Hazard: Irritant
Formula:
C₆H₁₁NO MW: 113.16 g/mol |
MDL Number:
MFCD00006483 CAS Number: 2591-86-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...