(3,4-Dimethoxyphenyl)acetic acid, Technical Grade
Supplier: Matrix Scientific
Synonyms:
Homoveratric acid
, 3,4-Dimethoxyphenylacetic acid
058301-25G
058301-100G
102551-520EA
64.62
USD
102551-520
102551-518
(3,4-Dimethoxyphenyl)acetic acid, Technical Grade
(3,4-Dimethoxyphenyl)acetic acid
Hazard: Irritant
Formula:
(C₆H₃(OCH₃)₂)CH₂COOH MW: 196.2 g/mol |
MDL Number:
MFCD00004335 CAS Number: 93-40-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...