3-(4-Bromophenyl)propan-1-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(4-Bromophenyl)-1-propanol3-(4-Bromophenyl)propyl alcohol

057601-5G 057601-25G
102551-484EA 752.31 USD
102551-484 102551-482
3-(4-Bromophenyl)propan-1-ol ≥95%
3-(4-Bromophenyl)propan-1-ol

Hazard: Irritant

Formula: C₉H₁₁BrO
MW: 215.09 g/mol
MDL Number: MFCD09028724
CAS Number: 25574-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR