MATR057253

057253-25G
102551-458EA 18.31 USD
102551-458
MATR057253
Purity: 95%
CAS: 74844-91-0
MDL Number: MFCD00076981
Molecular Formula: C11H19NO5
Molecular Weight: 245,28
Hazard: Irritant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR