3-Bromobenzyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Bromo-3-(chloromethyl)benzene1-(Chloromethyl)-3-bromobenzeneα-Chloro-3-bromotoluene3-Bromo-α-chlorotoluene

056901-25G 056901-5G
102551-430EA 58.46 USD
102551-430 102551-432
3-Bromobenzyl chloride ≥98%
3-Bromobenzyl chloride

Hazard: Corrosive/Lachrymator

Formula: C₇H₆BrCl
MW: 205.48 g/mol
Boiling Pt: 119 °C (18 mmHg)
Density: 1.56
MDL Number: MFCD00040865
CAS Number: 932-77-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR