2-methylpyrimidine-5-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 2-Methylpyrimidine-5-carboxylic acid

053743-1G 053743-5G
102551-400EA 44.62 USD
102551-400 102551-402
2-methylpyrimidine-5-carboxylic acid
2-methylpyrimidine-5-carboxylic acid

Hazard: Irritant

Formula: C₆H₆N₂O₂
MW: 138.126 g/mol
Melting Pt: 203…205 °C
MDL Number: MFCD08235210
CAS Number: 5194-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR