Methyl azetidine-3-carboxylate hydrochloride
Supplier: Matrix Scientific
Synonyms:
Azetidine-3-carboxylic acid methyll ester hydrochloride
053722-5G
053722-1G
102663-736EA
44.62
USD
102663-736
102663-738
Methyl azetidine-3-carboxylate hydrochloride
Methyl azetidine-3-carboxylate hydrochloride
Hazard: Irritant
Formula:
C₅H₁₀ClNO₂ MW: 151.59 g/mol |
MDL Number:
MFCD01861758 CAS Number: 100202-39-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...