3-Methyl-1H-1,2,4-triazole ≥95%

Supplier: Matrix Scientific

048112-1G
102833-160EA 34.5 USD
102833-160
3-Methyl-1H-1,2,4-triazole ≥95%
3-Methyl-1H-1,2,4-triazole

Hazard: Irritant

Formula: C₃H₅N₃
MW: 83.0900 g/mol
Boiling Pt: 138 °C (8 Torr)
Melting Pt: 94…96 °C
MDL Number: MFCD00233931
CAS Number: 7170-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR