Cytarabine ≥95%
Supplier: Matrix Scientific
Synonyms:
Cytosine beta-D-Arabinofuranoside
, Cytosine arabinoside
, Arabinocytidine
, 4-Amino-1-beta-D-arabinofuranosyl-2(1H)-pyrimidinone
, 1-β-D-Arabinofuranosylcytosine
, 4-Amino-1-((2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one
, Ara-C
047887-25G
102833-152EA
98.46
USD
102833-152
Cytarabine ≥95%
Cytarabine
Purity:
CAS: 69-74-9
MDL Number: MFCD00066487
Molecular Formula: C9H13N3O5
Molecular Weight: 243,22
Hazard: Irritant
Formula:
C₉H₁₃N₃O₅ MW: 243.22 g/mol Melting Pt: 197…198 °C |
MDL Number:
MFCD00066487 CAS Number: 147-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...