2-Amino-4-bromopyrimidine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromopyrimidin-2-amine

046666-1G
102833-142EA 135.38 USD
102833-142
2-Amino-4-bromopyrimidine ≥95%
2-Amino-4-bromopyrimidine

Hazard: Irritant

Formula: C₄H₄BrN₃
MW: 174.00 g/mol
Melting Pt: >300 °C
MDL Number: MFCD09999168
CAS Number: 343926-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR