Ethyl-6-oxo-2-(trifluoromethyl)-1,4,5,6-tetrahydro-3-pyridinecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 6-oxo-2-(trifluoromethyl)-1,4,5,6-tetrahydropyridine-3-carboxylate
, Ethyl 6-oxo-1,4,5,6-tetrahydro-2-(trifluoromethyl)pyridine-3-carboxylate
043511-5G
043511-1G
102551-320EA
123.08
USD
102551-320
102551-322
Ethyl-6-oxo-2-(trifluoromethyl)-1,4,5,6-tetrahydro-3-pyridinecarboxylate ≥95%
Ethyl-6-oxo-2-(trifluoromethyl)-1,4,5,6-tetrahydro-3-pyridinecarboxylate
Hazard: Irritant
Formula:
C₉H₁₀F₃NO₃ MW: 237.178 g/mol Melting Pt: 128…130 °C |
MDL Number:
MFCD00975005 CAS Number: 194673-12-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...