(S)-(-)-2-Pyrrolidinone-5-carboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
(S)-(-)-2-Pyrrolidone-5-carboxylic acid
, L-Pyroglutamic Acid
, (S)-5-Oxopyrrolidine-2-carboxylic acid
, H-Pyr-OH
041659-500G
041659-100G
102551-292EA
63.08
USD
102551-292
102551-294
(S)-(-)-2-Pyrrolidinone-5-carboxylic acid ≥95%
(S)-(-)-2-Pyrrolidinone-5-carboxylic acid
Hazard: Irritant
Formula:
C₅H₇NO₃ MW: 129.1 g/mol |
MDL Number:
MFCD00005272 CAS Number: 98-79-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...