5-Chloro-1H-indole-3-carbaldehyde
Supplier: Matrix Scientific
Synonyms:
5-Chloroindole-3-carboxaldehyde
038725-5G
102551-264EA
429.23
USD
102551-264
5-Chloro-1H-indole-3-carbaldehyde
5-Chloro-1H-indole-3-carbaldehyde
Hazard: Irritant
Formula:
C₉H₆ClNO MW: 179.61 g/mol |
MDL Number:
MFCD00175291 CAS Number: 827-01-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...