2,5-Dimercapto-1,3,4-thiadiazole
Supplier: Matrix Scientific
Synonyms:
1,3,4-Thiadiazole-2,5-dithiol
038238-100G
102551-254EA
47.69
USD
102551-254
2,5-Dimercapto-1,3,4-thiadiazole
2,5-Dimercapto-1,3,4-thiadiazole
Hazard: Irritant-Harmful/Refrigerate/Stench
Formula:
C₂H₂N₂S₃ MW: 150.25 g/mol Melting Pt: 161…164 °C. (dec.) |
MDL Number:
MFCD00003103 CAS Number: 1072-71-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...