β-Alanine-tert-butylester hydrochloride

Supplier: Matrix Scientific

Synonyms: H-β-Ala-OtBu·HClH-beta-Ala-OtBu.HClbeta-Alanine tert-Butyl ester hydrochlorideH-beta-Ala-OtBu·HClH-β-Ala-OtBu.HCl

037595-5G
102833-078EA 30.77 USD
102833-078
β-Alanine-tert-butylester hydrochloride
beta-Alanine-tert-butylester hydrochloride

Hazard: Irritant

Formula: C₇H₁₆ClNO₂
MW: 181.66 g/mol
MDL Number: MFCD00058258
CAS Number: 58620-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR