(±)-1-Aminoindan

Supplier: Matrix Scientific

Synonyms: 2,3-Dihydro-1H-inden-1-amine(±)-1-Indanylamine

037567-5G 037567-25G
102551-224EA 36.92 USD
102551-224 102551-226
(±)-1-Aminoindan
(±)-1-Aminoindan

Hazard: Irritant

Formula: C₉H₁₁N
MW: 133.19 g/mol
MDL Number: MFCD00003799
CAS Number: 34698-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR