2-Mercapto-5-methoxybenzimidazole

Supplier: Matrix Scientific

Synonyms: 5-Methoxy-1H-benzo[d]imidazole-2-thiol5-Methoxybenzimidazole-2-thiol2-Mercapto-5-methoxy-1H-benzimidazole5-Methoxy-2-benzimidazolethiol1,3-Dihydro-5-methoxy-2H-benzimidazole-2-thione

036092-100G 036092-25G
102551-178EA 501.95 USD
102551-178 102551-180
2-Mercapto-5-methoxybenzimidazole
2-Mercapto-5-methoxybenzimidazole

Hazard: Irritant

Formula: C₈H₈N₂OS
MW: 180.23 g/mol
MDL Number: MFCD00134581
CAS Number: 37052-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR