4-Vinylphenylboronic acid
Supplier: Matrix Scientific
Synonyms:
(4-Vinylphenyl)metaboric acid
, Dihydroxy(4-vinylphenyl)borane
, 4-Vinylbenzeneboronic acid
035623-25G
035623-5G
102833-040EA
316.92
USD
102833-040
102833-038
4-Vinylphenylboronic acid
4-Vinylphenylboronic acid
Hazard: Irritant/Refrigerate
Formula:
C₈H₉BO₂ MW: 147.97 g/mol Melting Pt: 179…192 °C |
MDL Number:
MFCD00239441 CAS Number: 2156-04-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...