4-Isopropoxyphenylboronic acid ≥98%
Supplier: Matrix Scientific
Synonyms:
4-Isopropoxybenzeneboronic acid
035611-25G
035611-100G
102663-682EA
110.77
USD
102663-682
102663-684
4-Isopropoxyphenylboronic acid ≥98%
4-Isopropoxyphenylboronic acid
Hazard: Irritant
Formula:
C₉H₁₃BO₃ MW: 180.01 g/mol |
MDL Number:
MFCD03427051 CAS Number: 153624-46-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...