4-Isobutylphenylboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Isobutylbenzeneboronic acid

035608-5G
102518-828EA 60 USD
102518-828
4-Isobutylphenylboronic acid ≥95%
4-Isobutylphenylboronic acid

Hazard: Irritant/Refrigerate

Formula: C₁₀H₁₅BO₂
MW: 178.04 g/mol
MDL Number: MFCD01632198
CAS Number: 153624-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR