4-(tert-Butoxycarboxy)phenylboronic acid

Supplier: Matrix Scientific

035546-1G 035546-5G
102518-820EA 138.46 USD
102518-820 102518-822
4-(tert-Butoxycarboxy)phenylboronic acid
4-(tert-Butoxycarboxy)phenylboronic acid

Hazard: Irritant/Refrigerate

Formula: C₁₁H₁₅BO₅
MW: 238.05 g/mol
MDL Number: MFCD03095144
CAS Number: 380430-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR