(3-Fluoro-4-hydroxyphenyl)boronic acid
Supplier: Matrix Scientific
Synonyms:
3-Fluoro-4-hydroxyphenylboronic acid
, 3-Fluoro-4-hydroxybenzeneboronic acid
035525-1G
102833-028EA
250.77
USD
102833-028
(3-Fluoro-4-hydroxyphenyl)boronic acid
(3-Fluoro-4-hydroxyphenyl)boronic acid
Hazard: Irritant
Formula:
C₅H₅BFNO₂ MW: 155.92 g/mol |
MDL Number:
MFCD06659838 CAS Number: 182344-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...