2,6-Dichloro-3-pyridineboronic acid

Supplier: Matrix Scientific

Synonyms: 2,6-Dichloropyridin-3-ylboronic acid2,6-Dichloropyridine-3-boronic acid

035318-1G
102833-016EA 93.85 USD
102833-016
2,6-Dichloro-3-pyridineboronic acid
2,6-Dichloro-3-pyridineboronic acid

Hazard: Irritant

Formula: C₅H₄BCl₂NO₂
MW: 191.81 g/mol
MDL Number: MFCD03094989
CAS Number: 148493-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR