2-Amino-5-bromo-3-methoxypyridine
Supplier: Matrix Scientific
Synonyms:
5-Bromo-3-methoxypyridin-2-amine
035155-1G
102551-168EA
210.77
USD
102551-168
2-Amino-5-bromo-3-methoxypyridine
2-Amino-5-bromo-3-methoxypyridine
Hazard: Irritant
Formula:
C₆H₇BrN₂O MW: 203.04 g/mol |
MDL Number:
MFCD09909849 CAS Number: 42409-58-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...