2-Amino-3-iodo-5-methylpyridine

Supplier: Matrix Scientific

Synonyms: 3-Iodo-5-methyl-2-pyridylamine2-Amino-3-iodo-5-picoline3-Iodo-5-methylpyridin-2-amine

035135-500MG
102551-166EA 230.77 USD
102551-166
2-Amino-3-iodo-5-methylpyridine
2-Amino-3-iodo-5-methylpyridine

Hazard: Irritant

Formula: C₆H₇IN₂
MW: 234.04 g/mol
MDL Number: MFCD09037445
CAS Number: 211308-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR