Diphenylacetic acid

Supplier: Matrix Scientific

Synonyms: 2,2-Diphenylacetic acid

031214-100G 031214-500G
102832-982EA 56.92 USD
102832-982 102832-984
Diphenylacetic acid
Diphenylacetic acid

Hazard: Irritant

Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
MDL Number: MFCD00004251
CAS Number: 117-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR