4-Amino-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

031135-1G 031135-5G
102663-662EA 126.15 USD
102663-662 102663-664
4-Amino-4H-1,2,4-triazole-3-thiol
4-Amino-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₂H₄N₄S
MDL Number: MFCD00128280
CAS Number: 4343-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR