(4-Chlorophenoxy)acetic acid

Supplier: Matrix Scientific

Synonyms: 4-CPA

031002-100G
102518-804EA 60 USD
102518-804
(4-Chlorophenoxy)acetic acid
(4-Chlorophenoxy)acetic acid

Hazard: Irritant

Formula: (C₆H₄Cl)OCH₂COOH
MW: 186.59 g/mol
MDL Number: MFCD00004305
CAS Number: 122-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR