3-Methylbutanohydrazide

Supplier: Matrix Scientific

029663-1G
102832-974EA 273.85 USD
102832-974
3-Methylbutanohydrazide
3-Methylbutanohydrazide

Hazard: Irritant

Formula: C₅H₁₂N₂O
MDL Number: MFCD00466383
CAS Number: 24310-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR