2-Chloro-N-(4-ethoxyphenyl)acetamide

Supplier: Matrix Scientific

029484-1G
102551-126EA 55.38 USD
102551-126
2-Chloro-N-(4-ethoxyphenyl)acetamide
2-Chloro-N-(4-ethoxyphenyl)acetamide

Hazard: Irritant

Formula: C₁₀H₁₂ClNO₂
Melting Pt: 145 °C
MDL Number: MFCD00018908
CAS Number: 2153-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR