N-(2-Aminoethyl)-N-(4-methoxyphenyl)amine

Supplier: Matrix Scientific

027328-1G
102832-962EA 263.08 USD
102832-962
N-(2-Aminoethyl)-N-(4-methoxyphenyl)amine
N-(2-Aminoethyl)-N-(4-methoxyphenyl)amine

Hazard: Irritant

Formula: C₉H₁₄N₂O
MDL Number: MFCD06795942
CAS Number: 24455-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR