1-(Pyridin-4-ylmethyl)piperazine
Supplier: Matrix Scientific
027302-5G
102551-112EA
592.31
USD
102551-112
1-(Pyridin-4-ylmethyl)piperazine
1-(Pyridin-4-ylmethyl)piperazine
Hazard: Irritant
Formula:
C₁₀H₁₅N₃ |
MDL Number:
MFCD01320892 CAS Number: 62089-74-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...