2-(4-Piperidinyl)benzoxazole
Supplier: Matrix Scientific
027245-5G
027245-1G
102832-956EA
167.69
USD
102832-956
102832-958
2-(4-Piperidinyl)benzoxazole
2-(4-Piperidinyl)benzoxazole
Hazard: Irritant
Formula:
C₁₂H₁₄N₂O |
MDL Number:
MFCD01475844 CAS Number: 51784-03-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...