Thiomorpholine-1,1-dioxide hydrochloride

Supplier: Matrix Scientific

Synonyms: Thiomorpholine 1,1-dioxide hydrochloride

027098-5G 027098-25G
102832-946EA 64.62 USD
102832-946 102832-948
Thiomorpholine-1,1-dioxide hydrochloride
Thiomorpholine 1,1-dioxide hydrochloride

Hazard: Irritant

Formula: C₄H₁₀ClNO₂S
MW: 171.6457 g/mol
MDL Number: MFCD06738995
CAS Number: 59801-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR