1H-Pyrrolo[2,3-b]pyridin-4-ol hydrate

Synonyms: 4-Hydroxy-7-azaindole hydrate1,7-dihydropyrrolo[2,3-b]pyridin-4-one

026232-1G
102551-088EA 794.86 USD
102551-088
1H-Pyrrolo[2,3-b]pyridin-4-ol hydrate
1H-Pyrrolo[2,3-b]pyridin-4-ol hydrate

Hazard: Irritant

Formula: C₇H₆N₂O·H₂O
MDL Number: MFCD08272229
CAS Number: 74420-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR