1H-Pyrrolo[2,3-b]pyridine-5-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 7-Azaindole-5-carboxylic acid

026198-250MG 026198-1G
102551-082EA 47.69 USD
102551-082 102551-084
1H-Pyrrolo[2,3-b]pyridine-5-carboxylic acid ≥95%
1H-Pyrrolo[2,3-b]pyridine-5-carboxylic acid

Hazard: Irritant

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
MDL Number: MFCD08272240
CAS Number: 754214-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR