5-Bromo-2-chloro-3-hydroxypyridine
Supplier: Matrix Scientific
Synonyms:
5-Bromo-2-chloropyridin-3-ol
026176-5G
026176-1G
102832-936EA
429.23
USD
102832-936
102832-938
5-Bromo-2-chloro-3-hydroxypyridine
5-Bromo-2-chloro-3-hydroxypyridine
Hazard: Irritant
Formula:
C₅H₃BrClNO MW: 208.4400 g/mol |
MDL Number:
MFCD08688592 CAS Number: 286946-77-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...