Ethyl-2-bromo-2-(3-chlorophen-1-yl)acetate ≥95%

Supplier: Matrix Scientific

024963-1G
102832-926EA 300 USD
102832-926
Ethyl-2-bromo-2-(3-chlorophen-1-yl)acetate ≥95%
Ethyl-2-bromo-2-(3-chlorophen-1-yl)acetate

Hazard: Irritant

Formula: C₁₀H₁₀BrClO₂
MW: 277.55 g/mol
MDL Number: MFCD04116364
CAS Number: 41024-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR