4-Phenoxypiperidine

Supplier: Matrix Scientific

023265-1G
102663-638EA 235.75 USD
102663-638
4-Phenoxypiperidine
4-Phenoxypiperidine

Hazard: Irritant

Formula: C₁₁H₁₅NO
MW: 177.25 g/mol
MDL Number: MFCD04114971
CAS Number: 3202-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR