(2R,3R)-2-(3-Chlorophenyl)-1-ethyl-6-oxopiperidine-3-carboxylic acid
Supplier: Matrix Scientific
021605-1G
102465-372EA
581.54
USD
102465-372
(2R,3R)-2-(3-Chlorophenyl)-1-ethyl-6-oxopiperidine-3-carboxylic acid
(2R,3R)-2-(3-Chlorophenyl)-1-ethyl-6-oxopiperidine-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₄H₁₆ClNO₃ |
MDL Number:
MFCD08687455 CAS Number: 1175689-25-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...