3,5-Dimethylisoxazole-4-sulfonyl chloride
Supplier: Matrix Scientific
021480-1G
021480-5G
102551-024EA
80
USD
102551-024
102551-026
3,5-Dimethylisoxazole-4-sulfonyl chloride
3,5-Dimethylisoxazole-4-sulphonyl chloride
Hazard: Corrosive
Formula:
C₅H₆ClNO₃S MW: 195.62 g/mol Melting Pt: 37…40 °C |
MDL Number:
MFCD00052554 CAS Number: 80466-79-1 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...