1H-Pyrazole-3-boronic acid

Supplier: Matrix Scientific

Synonyms: (1H-Pyrazol-3-yl)boronic acid

021354-1G
102663-632EA 150.77 USD
102663-632
1H-Pyrazole-3-boronic acid
1H-Pyrazole-3-boronic acid

Hazard: Irritant

Formula: C₃H₅BN₂O₂
MW: 111.89 g/mol
MDL Number: MFCD02020768
CAS Number: 376584-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR