2-Difluoromethyl-1H-benzoimidazole

Supplier: Matrix Scientific

Synonyms: 2-(Difluoromethyl)benzimidazole2-(Difluoromethyl)-1H-benzo[d]imidazole

021277-1G
102551-012EA 200 USD
102551-012
2-Difluoromethyl-1H-benzoimidazole
2-Difluoromethyl-1H-benzoimidazole

Hazard: Irritant

Formula: C₈H₆F₂N₂
MW: 168.15 g/mol
MDL Number: MFCD00463015
CAS Number: 705-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR