1H-Pyrrolo[2,3-b]pyridine-5-carbonitrile ≥98%

Supplier: Matrix Scientific

021145-1G
102551-006EA 129.23 USD
102551-006
1H-Pyrrolo[2,3-b]pyridine-5-carbonitrile ≥98%
1H-Pyrrolo[2,3-b]pyridine-5-carbonitrile

Hazard: Irritant

Formula: C₈H₅N₃
MW: 143.15 g/mol
Melting Pt: 225.1…225.2 °C
MDL Number: MFCD06659684
CAS Number: 517918-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR