5-Methylisoxazole-4-carbonyl chloride
Supplier: Matrix Scientific
019946-1G
102551-000EA
321.54
USD
102551-000
5-Methylisoxazole-4-carbonyl chloride
5-Methylisoxazole-4-carbonyl chloride
Hazard: Irritant
Formula:
C₅H₄ClNO₂ MW: 145.55 g/mol |
MDL Number:
MFCD03411599 CAS Number: 67305-24-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...