1-Benzylazetidine-3-carboxylic acid

019428-5G
102832-888EA 603.75 USD
102832-888
1-Benzylazetidine-3-carboxylic acid
1-Benzylazetidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₃NO₂
MDL Number: MFCD06752277
CAS Number: 94985-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR