3-Phenylpyrrolidine

Supplier: Matrix Scientific

018619-1G
102663-612EA 183.08 USD
102663-612
3-Phenylpyrrolidine
3-Phenylpyrrolidine

Hazard: Irritant

Formula: C₁₀H₁₃N
MW: 147.22 g/mol
MDL Number: MFCD01838169
CAS Number: 936-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR