2-Bromo-N-methylacetamide
Supplier: Matrix Scientific
017248-1G
102550-990EA
284.62
USD
102550-990
2-Bromo-N-methylacetamide
2-Bromo-N-methylacetamide
Hazard: Irritant
Formula:
C₃H₆BrNO |
MDL Number:
MFCD06800288 CAS Number: 34680-81-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...