[(1-Benzylpiperidin-4-yl)methyl]amine
Supplier: Matrix Scientific
016441-1G
102550-988EA
255.38
USD
102550-988
[(1-Benzylpiperidin-4-yl)methyl]amine
[(1-Benzylpiperidin-4-yl)methyl]amine
Hazard: Irritant
Formula:
C₁₃H₂₀N₂ |
MDL Number:
MFCD04116211 CAS Number: 88915-26-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...